siRNA Details
| Gene Symbol: Tgfbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012775.2 | NP_036907.2 | 174 | |||
| XM_006238030.2 | XP_006238092.2 | 176 | ||||
| GenBank mRNA | BC087035.1 | - | 116 | 7098473 | ||
| CB804748.1 | - | 218 | ||||
| L26110.1 | - | 131 | ||||

Tgfbr1
Genomic Map