siRNA Details
| Gene Symbol: Dlg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012788.1 | NP_036920.1 | 3, 4 | 600 | ||
| GenBank mRNA | U14950.1 | - | 3, 4 | 600 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlg1| Gene Symbol: Dlg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012788.1 | NP_036920.1 | 3, 4 | 600 | ||
| GenBank mRNA | U14950.1 | - | 3, 4 | 600 | ||