siRNA Details
| Gene Symbol: Mapk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270556.1 | NP_001257485.1 | 4, 5 | 745 | ||
| NM_012806.2 | NP_036938.2 | 4, 5 | 772 | |||
| XM_008769992.1 | XP_008768214.1 | 3, 4 | 3564 | |||
| GenBank mRNA | DQ377224.1 | - | 1, 2 | 119 | ||
| FQ211777.1 | - | 4, 5 | 744 | |||
| L27128.1 | - | 4, 5 | 482 | |||

Mapk10
Genomic Map