siRNA Details
| Gene Symbol: Prkcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M13706.1 | - | 337 | |||
| M19007.1 | - | 1, 2 | 287 | |||
| X04440.1 | - | 881 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkcb| Gene Symbol: Prkcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M13706.1 | - | 337 | |||
| M19007.1 | - | 1, 2 | 287 | |||
| X04440.1 | - | 881 | ||||