siRNA Details
| Gene Symbol: Lipa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012732.3 | NP_036864.3 | 3, 4 | 517 | ||
| XM_006231275.2 | XP_006231337.1 | 3, 4 | 484 | |||
| XM_008760321.1 | XP_008758543.1 | 2, 3 | 380 | |||
| GenBank mRNA | BC072532.1 | - | 3, 4 | 517 | 7099460 | |
| S81497.1 | - | 3, 4 | 457 | |||

Lipa
Genomic Map