siRNA Details
| Gene Symbol: Hk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012735.2 | NP_036867.1 | 7 | 924 | ||
| XM_006236711.2 | XP_006236773.1 | 6 | 1993 | |||
| GenBank mRNA | AF027179.1 | - | 7 | 728 | ||
| M68971.1 | - | 7 | 925 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hk2| Gene Symbol: Hk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012735.2 | NP_036867.1 | 7 | 924 | ||
| XM_006236711.2 | XP_006236773.1 | 6 | 1993 | |||
| GenBank mRNA | AF027179.1 | - | 7 | 728 | ||
| M68971.1 | - | 7 | 925 | |||