siRNA Details
| Gene Symbol: Slc2a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012751.1 | NP_036883.1 | 3, 4 | 471 | ||
| XM_006246596.2 | XP_006246658.1 | 4, 5 | 444 | |||
| GenBank mRNA | BC085757.1 | - | 3, 4 | 516 | 7192278 | |
| D28561.1 | - | 3, 4 | 471 | |||
| FQ216450.1 | - | 4, 5 | 410 | |||
| FQ232657.1 | - | 3, 4 | 530 | |||
| FQ233419.1 | - | 3, 4 | 506 | |||
| J04524.1 | - | 3, 4 | 453 | |||
| KJ696745.1 | - | 3, 4 | 356 | |||
| M25482.1 | - | 3, 4 | 436 | |||
| X14771.1 | - | 3, 4 | 484 | |||

Slc2a4
Genomic Map