siRNA Details
| Gene Symbol: Dyrk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012791.2 | NP_036923.1 | 3, 4 | 342 | ||
| XM_006248031.2 | XP_006248093.1 | 3, 4 | 3287 | |||
| XM_008768562.1 | XP_008766784.1 | 2, 3 | 2550 | |||
| XM_008768563.1 | XP_008766785.1 | 4, 5 | 3423 | |||
| XM_008768564.1 | XP_008766786.1 | 3, 4 | 389 | |||
| GenBank mRNA | X79769.1 | - | 3, 4 | 344 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001113389, NM_007890, BC138716, U58497, CD540384, and others.

Dyrk1a
Genomic Map