siRNA Details
| Gene Symbol: P2rx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012721.2 | NP_036853.2 | 12 | 1259 | ||
| XM_008768851.1 | XP_008767073.1 | 11 | 1530 | |||
| XM_008768852.1 | XP_008767074.1 | 11 | 1215 | |||
| GenBank mRNA | CO554543.1 | - | 2 | 110 | ||
| X92070.1 | - | 12 | 1140 | |||
| X97376.1 | - | 12 | 1270 | |||

P2rx6
Genomic Map