siRNA Details
| Gene Symbol: Camk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012727.3 | NP_036859.2 | 13 | 1529 | ||
| XM_006254523.2 | XP_006254585.1 | 12 | 1471 | |||
| GenBank mRNA | BC128706.1 | - | 13 | 1529 | 7455982 | |
| J04446.1 | - | 3 | 604 | |||
| J04600.1 | - | 556 | ||||
| M63334.1 | - | 1423 | ||||
| M64757.1 | - | 11 | 1519 | |||
| S65840.1 | - | 12 | 1454 | |||

Camk4
Genomic Map