siRNA Details
| Gene Symbol: Pc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012744.2 | NP_036876.2 | 9 | 1166 | ||
| XM_006230689.2 | XP_006230751.1 | 10 | 1397 | |||
| XM_006230690.2 | XP_006230752.1 | 9 | 1512 | |||
| XM_006230691.2 | XP_006230753.1 | 10 | 1219 | |||
| XM_006230692.2 | XP_006230754.1 | 9 | 1189 | |||
| XM_006230693.2 | XP_006230755.1 | 10 | 1321 | |||
| XM_006230694.2 | XP_006230756.1 | 9 | 1188 | |||
| XM_008760087.1 | XP_008758309.1 | 9 | 1294 | |||
| GenBank mRNA | BC085680.1 | - | 9 | 1143 | 7109369 | |
| U32314.1 | - | 9 | 1134 | |||
| U36585.1 | - | 9 | 1166 | |||

Pc
Genomic Map