siRNA Details
| Gene Symbol: Gfra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012750.1 | NP_036882.1 | 9 | 1433 | ||
| XM_006252238.2 | XP_006252300.1 | 8 | 2202 | |||
| GenBank mRNA | AF003825.1 | - | 1983 | |||
| AF005226.1 | - | 9 | 1313 | |||
| U97143.1 | - | 9 | 1433 | |||

Gfra2
Genomic Map