siRNA Details
| Gene Symbol: Aqp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012778.1 | NP_036910.1 | 4 | 821 | ||
| GenBank mRNA | BC090068.1 | - | 4 | 803 | 7190406 | |
| L07268.1 | - | 4 | 812 | |||
| X67948.1 | - | 4 | 821 | |||
| X70257.1 | - | 4 | 761 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aqp1| Gene Symbol: Aqp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012778.1 | NP_036910.1 | 4 | 821 | ||
| GenBank mRNA | BC090068.1 | - | 4 | 803 | 7190406 | |
| L07268.1 | - | 4 | 812 | |||
| X67948.1 | - | 4 | 821 | |||
| X70257.1 | - | 4 | 761 | |||