siRNA Details
| Gene Symbol: St8sia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012813.2 | NP_036945.2 | 1 | 470 | ||
| XM_008763368.1 | XP_008761590.1 | 1 | 470 | |||
| GenBank mRNA | CB556962.1 | - | 1 | 474 | ||
| D45255.1 | - | 1 | 221 | |||
| U53883.1 | - | 1 | 191 | |||

St8sia1
Genomic Map