siRNA Details
| Gene Symbol: Acadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012819.1 | NP_036951.1 | 3 | 303 | ||
| GenBank mRNA | BC062006.1 | - | 3 | 301 | 5599828 | |
| FQ215575.1 | - | 3 | 354 | |||
| J05029.1 | - | 3 | 314 | |||
| L11276.1 | - | 3 | 303 | |||

Acadl
Genomic Map