siRNA Details
| Gene Symbol: Aqp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142366.2 | NP_001135838.1 | 5 | 815 | ||
| NM_001270558.2 | NP_001257487.1 | 4 | 642 | |||
| NM_001270559.2 | NP_001257488.1 | 4 | 650 | |||
| NM_012825.4 | NP_036957.1 | 5 | 807 | |||
| GenBank mRNA | L27588.1 | - | 4 | 675 | ||
| U14007.1 | - | 5 | 741 | |||

Aqp4
Genomic Map