siRNA Details
| Gene Symbol: Azgp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D21058.1 | - | 2 | 137 | ||
| X75309.1 | - | 2 | 119 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Azgp1| Gene Symbol: Azgp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D21058.1 | - | 2 | 137 | ||
| X75309.1 | - | 2 | 119 | |||