siRNA Details
| Gene Symbol: Cacnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012828.2 | NP_036960.1 | 3 | 291 | ||
| XM_006257329.2 | XP_006257391.1 | 3 | 363 | |||
| XM_006257330.2 | XP_006257392.1 | 3 | 772 | |||
| GenBank mRNA | CB739910.1 | - | 3 | 417 | ||
| M88751.1 | - | 3 | 295 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001286226, NM_001044741, NM_007581, U20372, AK197162, and others.

Cacnb3
Genomic Map