siRNA Details
Gene Symbol: Ms4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012845.1 | NP_036977.1 | 4 | 421 | ||
GenBank mRNA | M22923.1 | - | 4 | 421 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ms4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012845.1 | NP_036977.1 | 4 | 421 | ||
GenBank mRNA | M22923.1 | - | 4 | 421 |