siRNA Details
| Gene Symbol: Fnta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012847.1 | NP_036979.1 | 4 | 489 | ||
| GenBank mRNA | BC085758.1 | - | 4 | 439 | 7189836 | |
| M81225.1 | - | 4 | 489 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fnta| Gene Symbol: Fnta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012847.1 | NP_036979.1 | 4 | 489 | ||
| GenBank mRNA | BC085758.1 | - | 4 | 439 | 7189836 | |
| M81225.1 | - | 4 | 489 | |||