siRNA Details
Gene Symbol: Mmp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012864.2 | NP_036996.1 | 1, 2 | 138 | ||
GenBank mRNA | BC064657.1 | - | 1, 2 | 138 | 5623517 | |
L24374.1 | - | 1, 2 | 152 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mmp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012864.2 | NP_036996.1 | 1, 2 | 138 | ||
GenBank mRNA | BC064657.1 | - | 1, 2 | 138 | 5623517 | |
L24374.1 | - | 1, 2 | 152 |