siRNA Details
| Gene Symbol: Nfyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012866.2 | NP_036998.1 | 4 | 417 | ||
| XM_006238785.2 | XP_006238847.1 | 4 | 390 | |||
| GenBank mRNA | BC093619.1 | - | 4 | 417 | 7314492 | |
| U17607.1 | - | 4 | 305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfyc| Gene Symbol: Nfyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012866.2 | NP_036998.1 | 4 | 417 | ||
| XM_006238785.2 | XP_006238847.1 | 4 | 390 | |||
| GenBank mRNA | BC093619.1 | - | 4 | 417 | 7314492 | |
| U17607.1 | - | 4 | 305 | |||