siRNA Details
Gene Symbol: Pdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012872.1 | NP_037004.1 | 3 | 159 | ||
GenBank mRNA | M33528.1 | - | 3 | 159 | ||
M33530.1 | - | 3 | 169 | |||
M60738.1 | - | 2 | 132 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012872.1 | NP_037004.1 | 3 | 159 | ||
GenBank mRNA | M33528.1 | - | 3 | 159 | ||
M33530.1 | - | 3 | 169 | |||
M60738.1 | - | 2 | 132 |