siRNA Details
| Gene Symbol: Rps29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012876.1 | NP_037008.1 | 2 | 189 | ||
| GenBank mRNA | BC058150.1 | - | 2 | 187 | 6922365 | |
| FQ209501.1 | - | 2 | 183 | |||
| FQ210507.1 | - | 2 | 183 | |||
| FQ211740.1 | - | 2 | 183 | |||
| FQ216880.1 | - | 2 | 183 | |||
| FQ217140.1 | - | 2 | 183 | |||
| FQ217213.1 | - | 2 | 183 | |||
| FQ217476.1 | - | 2 | 183 | |||
| FQ218006.1 | - | 2 | 183 | |||
| FQ218037.1 | - | 2 | 184 | |||
| FQ218769.1 | - | 2 | 183 | |||
| FQ221030.1 | - | 2 | 183 | |||
| FQ221251.1 | - | 2 | 183 | |||
| FQ221298.1 | - | 2 | 183 | |||
| FQ221307.1 | - | 2 | 180 | |||
| FQ221360.1 | - | 2 | 183 | |||
| FQ221457.1 | - | 2 | 183 | |||
| FQ221523.1 | - | 2 | 183 | |||
| FQ221587.1 | - | 2 | 184 | |||
| FQ221640.1 | - | 2 | 183 | |||
| FQ221661.1 | - | 2 | 183 | |||
| FQ221707.1 | - | 2 | 183 | |||
| FQ221875.1 | - | 2 | 183 | |||
| FQ221942.1 | - | 2 | 183 | |||
| FQ222143.1 | - | 2 | 183 | |||
| FQ222293.1 | - | 2 | 183 | |||
| FQ222368.1 | - | 2 | 183 | |||
| FQ222566.1 | - | 2 | 183 | |||
| FQ222711.1 | - | 2 | 183 | |||
| FQ222937.1 | - | 2 | 183 | |||
| FQ223071.1 | - | 2 | 183 | |||
| FQ223222.1 | - | 2 | 183 | |||
| FQ223289.1 | - | 2 | 183 | |||
| FQ223378.1 | - | 2 | 183 | |||
| FQ223402.1 | - | 2 | 183 | |||
| FQ223885.1 | - | 2 | 183 | |||
| FQ223975.1 | - | 2 | 183 | |||
| FQ224325.1 | - | 2 | 183 | |||
| FQ224400.1 | - | 2 | 183 | |||
| FQ224465.1 | - | 2 | 183 | |||
| FQ224476.1 | - | 2 | 183 | |||
| FQ224661.1 | - | 2 | 184 | |||
| FQ224820.1 | - | 2 | 183 | |||
| FQ228434.1 | - | 2 | 183 | |||
| FQ228730.1 | - | 2 | 183 | |||
| X59051.1 | - | 2 | 189 | |||

Rps29
Genomic Map