siRNA Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012891.2 | NP_037023.1 | 2, 3 | 165 | ||
| GenBank mRNA | BC087036.1 | - | 2, 3 | 186 | 7193535 | |
| D30647.1 | - | 2, 3 | 167 | |||
| FQ210754.1 | - | 2, 3 | 214 | |||
| FQ219329.1 | - | 2, 3 | 210 | |||

Acadvl
Genomic Map