siRNA Details
| Gene Symbol: Alad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012899.2 | NP_037031.1 | 3, 4 | 337 | ||
| XM_006238234.2 | XP_006238296.1 | 3, 4 | 930 | |||
| XM_006238235.1 | XP_006238297.1 | 3, 4 | 427 | |||
| GenBank mRNA | BC061806.1 | - | 3, 4 | 337 | 5598340 | |
| FQ210250.1 | - | 3, 4 | 282 | |||
| FQ213585.1 | - | 3, 4 | 479 | |||
| FQ218546.1 | - | 3, 4 | 282 | |||
| FQ219571.1 | - | 3, 4 | 282 | |||
| FQ231154.1 | - | 3, 4 | 313 | |||
| FQ232546.1 | - | 3, 4 | 313 | |||
| FQ233724.1 | - | 3, 4 | 363 | |||
| FQ234573.1 | - | 3, 4 | 361 | |||
| X04959.1 | - | 2, 3 | 219 | |||

Alad
Genomic Map