siRNA Details
| Gene Symbol: Ambn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012900.1 | NP_037032.1 | 5 | 388 | ||
| GenBank mRNA | U35097.1 | - | 5 | 388 | ||
| Z50083.1 | - | 5 | 391 | |||
| Z50084.1 | - | 1 | 54 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ambn| Gene Symbol: Ambn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012900.1 | NP_037032.1 | 5 | 388 | ||
| GenBank mRNA | U35097.1 | - | 5 | 388 | ||
| Z50083.1 | - | 5 | 391 | |||
| Z50084.1 | - | 1 | 54 | |||