siRNA Details
| Gene Symbol: Arrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012910.2 | NP_037042.1 | 4, 5 | 192 | ||
| XM_006229736.2 | XP_006229798.1 | 4, 5 | 787 | |||
| XM_006229737.2 | XP_006229799.1 | 4, 5 | 786 | |||
| GenBank mRNA | M91589.1 | - | 4, 5 | 195 | ||

Arrb1
Genomic Map