siRNA Details
| Gene Symbol: Atp2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012914.1 | NP_037046.1 | 4, 5 | 463 | ||
| XM_006246597.2 | XP_006246659.1 | 5, 6 | 1077 | |||
| GenBank mRNA | M30581.1 | - | 4, 5 | 463 | ||
| Gene Symbol: LOC102549389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768230.1 | XP_008766452.1 | 2 | 101 | ||

Atp2a3
Genomic Map