siRNA Details
| Gene Symbol: Bcan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033665.1 | NP_001028837.1 | 3 | 557 | ||
| NM_012916.2 | NP_037048.2 | 3 | 557 | |||
| XM_006232595.2 | XP_006232657.1 | 3 | 1638 | |||
| XM_006232596.2 | XP_006232658.1 | 3 | 541 | |||
| XM_006232597.2 | XP_006232659.1 | 3 | 359 | |||
| GenBank mRNA | BC094307.1 | - | 1 | 818 | ||
| U37142.1 | - | 3 | 361 | |||
| X79881.1 | - | 3 | 411 | |||
| X86406.1 | - | 3 | 411 | |||
| Z28366.1 | - | 3 | 552 | |||

Bcan
Genomic Map