siRNA Details
| Gene Symbol: DEK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003472.3 | NP_003463.1 | 3 | 367 | ||
| XM_017011273.1 | XP_016866762.1 | 3 | 339 | |||
| GenBank mRNA | AK294079.1 | - | 3 | 334 | ||
| AK298089.1 | - | 3 | 279 | |||
| AK312616.1 | - | 3 | 296 | |||
| AU131262.1 | - | 3 | 367 | |||
| BC035259.1 | - | 3 | 339 | 5122743 | ||
| X64229.1 | - | 3 | 207 | |||

DEK
Genomic Map