siRNA Details
Gene Symbol: Gclc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012815.2 | NP_036947.1 | 3 | 405 | ||
GenBank mRNA | BC081702.1 | - | 3 | 405 | 7110739 | |
J05181.1 | - | 3 | 377 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gclc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012815.2 | NP_036947.1 | 3 | 405 | ||
GenBank mRNA | BC081702.1 | - | 3 | 405 | 7110739 | |
J05181.1 | - | 3 | 377 |