siRNA Details
| Gene Symbol: Adcy6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270785.1 | NP_001257714.1 | 2 | 429 | ||
| GenBank mRNA | L01115.1 | - | 2 | 572 | ||
| M96160.1 | - | 2 | 429 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adcy6| Gene Symbol: Adcy6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270785.1 | NP_001257714.1 | 2 | 429 | ||
| GenBank mRNA | L01115.1 | - | 2 | 572 | ||
| M96160.1 | - | 2 | 429 | |||