siRNA Details
| Gene Symbol: Cstb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012838.2 | NP_036970.1 | 2, 3 | 273 | ||
| GenBank mRNA | BC084725.1 | - | 2, 3 | 261 | 7318993 | |
| CF976177.1 | - | 2, 3 | 274 | |||
| FQ220397.1 | - | 2, 3 | 257 | |||
| FQ221069.1 | - | 2, 3 | 233 | |||
| FQ222613.1 | - | 2, 3 | 228 | |||
| X54737.1 | - | 2, 3 | 265 | |||

Cstb
Genomic Map