siRNA Details
| Gene Symbol: Gast | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012849.1 | NP_036981.1 | 3 | 305 | ||
| GenBank mRNA | M38653.1 | - | 3 | 305 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gast| Gene Symbol: Gast | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012849.1 | NP_036981.1 | 3 | 305 | ||
| GenBank mRNA | M38653.1 | - | 3 | 305 | ||