siRNA Details
| Gene Symbol: Lipe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012859.1 | NP_036991.1 | 1 | 394 | ||
| GenBank mRNA | BC078888.1 | - | 1 | 377 | ||
| U40001.1 | - | 1 | 394 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lipe| Gene Symbol: Lipe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012859.1 | NP_036991.1 | 1 | 394 | ||
| GenBank mRNA | BC078888.1 | - | 1 | 377 | ||
| U40001.1 | - | 1 | 394 | |||