siRNA Details
| Gene Symbol: Tnfrsf11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012870.2 | NP_037002.1 | 3 | 641 | ||
| GenBank mRNA | BC081830.1 | - | 3 | 641 | 7096099 | |
| U94330.1 | - | 3 | 660 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfrsf11b| Gene Symbol: Tnfrsf11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012870.2 | NP_037002.1 | 3 | 641 | ||
| GenBank mRNA | BC081830.1 | - | 3 | 641 | 7096099 | |
| U94330.1 | - | 3 | 660 | |||