siRNA Details
| Gene Symbol: Spp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012881.2 | NP_037013.2 | 6 | 417 | ||
| XM_008769996.1 | XP_008768218.1 | 6 | 490 | |||
| GenBank mRNA | AB001382.1 | - | 5 | 311 | ||
| BC078874.1 | - | 6 | 397 | 7110677 | ||
| CV102664.1 | - | 6 | 417 | |||
| FQ213473.1 | - | 6 | 406 | |||
| FQ222192.1 | - | 6 | 433 | |||
| M14656.1 | - | 6 | 390 | |||
| M99252.1 | - | 6 | 426 | |||

Spp1
Genomic Map