siRNA Details
| Gene Symbol: Timp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012886.2 | NP_037018.2 | 4 | 695 | ||
| XM_006241162.2 | XP_006241224.1 | 4 | 724 | |||
| GenBank mRNA | BC097335.1 | - | 4 | 695 | 7443686 | |
| U27201.1 | - | 4 | 410 | |||

Timp3
Genomic Map