siRNA Details
| Gene Symbol: Adrbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012897.2 | NP_037029.1 | 3 | 257 | ||
| XM_008769317.1 | XP_008767539.1 | 2 | 148 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adrbk2| Gene Symbol: Adrbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012897.2 | NP_037029.1 | 3 | 257 | ||
| XM_008769317.1 | XP_008767539.1 | 2 | 148 | |||