siRNA Details
| Gene Symbol: Apobec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012907.2 | NP_037039.1 | 3 | 890 | ||
| XM_006237290.2 | XP_006237352.1 | 4 | 1082 | |||
| XM_006237291.2 | XP_006237353.1 | 4 | 1138 | |||
| XM_006237292.2 | XP_006237354.1 | 4 | 659 | |||
| XM_006237293.2 | XP_006237355.1 | 4 | 814 | |||
| XM_008763219.1 | XP_008761441.1 | 4 | 729 | |||
| XM_008763220.1 | XP_008761442.1 | 4 | 598 | |||
| GenBank mRNA | BC085335.1 | - | 3 | 890 | 7301703 | |
| L07114.1 | - | 3 | 426 | |||

Apobec1
Genomic Map