siRNA Details
| Gene Symbol: Acsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012820.1 | NP_036952.1 | 18, 19 | 1901 | ||
| XM_006253122.1 | XP_006253184.1 | 19, 20 | 2561 | |||
| XM_006253123.1 | XP_006253185.1 | 19, 20 | 2102 | |||
| XM_006253124.2 | XP_006253186.1 | 19, 20 | 2021 | |||
| XM_006253125.2 | XP_006253187.1 | 19, 20 | 2181 | |||
| XM_006253126.1 | XP_006253188.1 | 19, 20 | 2027 | |||
| XM_006253127.1 | XP_006253189.1 | 19, 20 | 1992 | |||
| XM_008771254.1 | XP_008769476.1 | 18, 19 | 1927 | |||
| GenBank mRNA | D90109.1 | - | 18, 19 | 1901 | ||
| FQ217275.1 | - | 18, 19 | 1871 | |||
| FQ219468.1 | - | 2, 3 | 202 | |||

Acsl1
Genomic Map