siRNA Details
| Gene Symbol: Aqp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142366.2 | NP_001135838.1 | 5 | 1021 | ||
| NM_001270558.2 | NP_001257487.1 | 4 | 848 | |||
| NM_001270559.2 | NP_001257488.1 | 4 | 856 | |||
| NM_012825.4 | NP_036957.1 | 5 | 1013 | |||
| GenBank mRNA | L27588.1 | - | 4 | 881 | ||
| U14007.1 | - | 5 | 947 | |||

Aqp4
Genomic Map