siRNA Details
| Gene Symbol: Abcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012833.2 | NP_036965.1 | 10, 11 | 1673 | ||
| GenBank mRNA | D86086.1 | - | 10, 11 | 1495 | ||
| L49379.1 | - | 10, 11 | 1554 | |||
| X96393.1 | - | 10, 11 | 1509 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcc2| Gene Symbol: Abcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012833.2 | NP_036965.1 | 10, 11 | 1673 | ||
| GenBank mRNA | D86086.1 | - | 10, 11 | 1495 | ||
| L49379.1 | - | 10, 11 | 1554 | |||
| X96393.1 | - | 10, 11 | 1509 | |||