siRNA Details
| Gene Symbol: Il10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012854.2 | NP_036986.2 | 3 | 327 | ||
| XM_006249712.2 | XP_006249774.1 | 3 | 666 | |||
| XM_008769426.1 | XP_008767648.1 | 3 | 666 | |||
| GenBank mRNA | L02926.1 | - | 3 | 327 | ||
| X60675.1 | - | 370 | ||||

Il10
Genomic Map