siRNA Details
| Gene Symbol: Mmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012864.2 | NP_036996.1 | 5 | 748 | ||
| GenBank mRNA | BC064657.1 | - | 5 | 748 | 5623517 | |
| L24374.1 | - | 5 | 762 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp7| Gene Symbol: Mmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012864.2 | NP_036996.1 | 5 | 748 | ||
| GenBank mRNA | BC064657.1 | - | 5 | 748 | 5623517 | |
| L24374.1 | - | 5 | 762 | |||