siRNA Details
| Gene Symbol: Nfya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012865.1 | NP_036997.1 | 4, 5 | 476 | ||
| XM_006244409.2 | XP_006244471.1 | 4, 5 | 483 | |||
| XM_006244410.1 | XP_006244472.1 | 4, 5 | 479 | |||
| XM_006244411.2 | XP_006244473.1 | 4, 5 | 473 | |||
| XM_006244412.2 | XP_006244474.1 | 4, 5 | 470 | |||
| XM_006244413.1 | XP_006244475.1 | 3, 4 | 391 | |||
| XM_008766839.1 | XP_008765061.1 | 3, 4 | 379 | |||
| GenBank mRNA | FQ210330.1 | - | 3, 4 | 396 | ||
| FQ223023.1 | - | 4, 5 | 488 | |||
| M34238.1 | - | 4, 5 | 476 | |||

Nfya
Genomic Map