siRNA Details
| Gene Symbol: Npr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012868.1 | NP_037000.1 | 7, 8 | 1520 | ||
| XM_006232045.2 | XP_006232107.1 | 7, 8 | 2107 | |||
| GenBank mRNA | L27339.1 | - | 7, 8 | 1520 | ||
| X75605.1 | - | 7, 8 | 1541 | |||
| X78595.1 | - | 1366 | ||||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001286395, NM_001039181, NM_008728, AK166535, AK079196, and others.

Npr3
Genomic Map