siRNA Details
| Gene Symbol: Tmpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012887.1 | NP_037019.1 | 8 | 1279 | ||
| XM_008765219.1 | XP_008763441.1 | 6 | 1102 | |||
| XM_008765220.1 | XP_008763442.1 | 5 | 991 | |||
| GenBank mRNA | U18314.1 | - | 8 | 1279 | ||

Tmpo
Genomic Map