siRNA Details
Gene Symbol: Vcam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012889.1 | NP_037021.1 | 1234 | |||
GenBank mRNA | BC089812.1 | - | 1252 | 7364929 | ||
M84488.1 | - | 1234 | ||||
X63722.1 | - | 1183 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vcam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012889.1 | NP_037021.1 | 1234 | |||
GenBank mRNA | BC089812.1 | - | 1252 | 7364929 | ||
M84488.1 | - | 1234 | ||||
X63722.1 | - | 1183 |